Learn More
Meso Scale Discovery U-PLEX Human RBP4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151V9K22
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for measuring retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a dynamic range and low sample volume The detection limit is 17 pg/mL and the quantification limit is 10 000 pg/mL It runs on MSD s SECTOR Imager 6000 for high throughput This assay accurately measures RBP4 linked to insulin resistance and obesity
Catalog No. 50-000-79078