Learn More
Meso Scale Discovery U-PLEX Human S100A12 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHPK21
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay kit that enables simultaneous quantification of S100A12 and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The assay has a typical lower limit of detection of 0 04 ng/mL for S100A12 and an upper limit of 10 000 pg/mL It allows high-throughput analysis of S100A12 levels
Catalog No. 50-000-80597