Learn More
Meso Scale Discovery U-PLEX Human TARC Assay, 384-well, Singleplex Assay, 0.51 pg/mL LLOD, Dynamic range 0.51-2200 pg/mL, Plasma, Serum, Cell Culture Supernatant
SDP
Supplier:  Meso Scale Discovery K251ULK2
Encompass
U-PLEX Human TARC Assay, 384-well, Singleplex Assay, 0.51 pg/mL LLOD, Dynamic range 0.51-2200 pg/mL, Plasma, Serum, Cell Culture Supernatant
Catalog No. NC2211652
Add to Cart
Request a Quote