Learn More
Meso Scale Discovery U-PLEX Human TARC ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151ULK4
Encompass
The Meso Scale Discovery U-PLEX Human TARC Assay is a multiplex immunoassay kit for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SECTOR Imager offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate capture and detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It quantifies TARC levels between 0 3-2500 pg/mL with minimal cross-reactivity enabling efficient and reproducible TARC biomarker analysis
Catalog No. 50-000-78242
Add to Cart
Request a Quote