Learn More
Meso Scale Discovery U-PLEX Human Tie-2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151F8K21
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Wash Buffer Diluent 100 and calibrators and controls to generate a standard curve and validate assay performance With a measuring time of 2 5 hours this kit allows quantification of human TIE-2 levels from 50-100 000 pg/mL
Catalog No. 50-000-81984