Learn More
Meso Scale Discovery U-PLEX Human TIGIT ASY
SDP
Supplier:  Meso Scale Discovery K151W7K2
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT Assay Kit is a multiplex immunoassay for quantifying human TIGIT and other proteins from serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents With a 3 5-hour protocol it efficiently profiles the human immunomodulatory proteome aiding in understanding immune regulation and response
Catalog No. 50-000-81860