Learn More
Meso Scale Discovery U-PLEX Human TLR1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151J7K22
Encompass
The U-PLEX Human TLR1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent This allows for the quantitative measurement of TLR1 levels from human samples With minimal hands-on time required this kit enables high-throughput screening and research on TLR1 signaling pathways
Catalog No. 50-000-79929