Learn More
Meso Scale Discovery U-PLEX Human TNF ASYQP
SDP
Supplier:  Meso Scale Discovery K151UCK21
Encompass
The U-PLEX Human TNF Assay (K151UCK) is a highly sensitive electrochemiluminescent immunoassay designed for quantitative measurement of tumor necrosis factor-alpha in human biological samples This multiplex-compatible assay utilizes MSD s proprietary U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards enabling researchers to accurately measure TNF levels in serum plasma and cell culture supernatants for inflammation and immune response studies
Catalog No. 50-000-79674