Learn More
Meso Scale Discovery U-PLEX Human TNF-RI ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHQK22
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF RI in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor soluble TNF RI levels as a biomarker and indicator of TNF pathway activation It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify samples
Catalog No. 50-000-80146