Learn More
Meso Scale Discovery U-PLEX Human TNF-RII Assay, detection of human TNF-RII, 1.6 - 7,000 pg/mL dynamic range, 96-well plate type, for research use only

Supplier: Meso Scale Discovery K151AHRK2
The MESO SCALE DISCOVERY U-PLEX Human TNF-RII Assay is a highly sensitive and flexible immunoassay designed for accurately detecting human Tumor Necrosis Factor Receptor II (TNF-RII). This assay offers validated components, ensuring reliable biomarker detection across various sample types. It supports multiplexing with Immuno-Oncology analytes and is intended for research use only.
- Assay platform is U-PLEX
- Analyte targets human TNF-RII
- Species reactivity is human
- Detection format is single-analyte, compatible with multiplex
- Uses a 96-well plate type
- Compatible with specific MESO instruments
- Sensitivity threshold is 1.6 pg/mL
- Dynamic range from 1.6 to 7,000 pg/mL
- Compatible sample types include plasma and serum
- Mouse monoclonal used for both capture and detection antibodies
- Calibrator type is recombinant human TNF-RII
- Nonspecific binding is less than 2.0%
- Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.