Learn More
Meso Scale Discovery U-PLEX Human TNF-RII ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHRK21
Encompass
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF receptor II (TNFRII) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance It can monitor soluble TNFRII levels and has applications in immunology oncology and drug development research
Catalog No. 50-000-80491