Learn More
Meso Scale Discovery U-PLEX Human TPO ASY
SDP
Supplier:  Meso Scale Discovery K151VKK2
Encompass
The Meso Scale Discovery U-PLEX Human TPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Thyroid Peroxidase (TPO) in human serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of 1000 ng/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay can aid in the diagnosis of autoimmune thyroid diseases
Catalog No. 50-000-81425