Learn More
Meso Scale Discovery U-PLEX Human TPO ASYQP
SDP
Supplier:  Meso Scale Discovery K151VKK22
Encompass
The MSD U-PLEX Human TPO Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of thrombopoietin (TPO) in human biological samples This singleplex assay utilizes MSD s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The assay is designed for research use in studying hematopoiesis platelet regulation and related disease states where TPO levels are clinically relevant
Catalog No. 50-000-81469