Learn More
Meso Scale Discovery U-PLEX Human TRAIL ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151XTK4
Encompass
The U-PLEX Human TRAIL Assay K151XTK is a multiplex immunoassay kit for the quantitative measurement of TRAIL in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay allows high sensitivity quantification of TRAIL across a wide dynamic range of 15 6 - 1000 pg/mL The assay requires a low sample volume of 50 L and provides results in under 3 hours This product enables researchers to accurately measure TRAIL levels for the study of apoptosis and immune system function
Catalog No. 50-000-80461
Add to Cart
Request a Quote