Learn More
Meso Scale Discovery U-PLEX Human TRAIL ASYQP
SDP
Supplier:  Meso Scale Discovery K151XTK22
Encompass
The U-PLEX Human TRAIL Assay (K151XTK) is a multiplex immunoassay designed to measure TRAIL (TNF-related apoptosis-inducing ligand) levels in human samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology for sensitive and specific quantification of TRAIL protein which plays a crucial role in apoptosis and immune system regulation The assay is suitable for research applications studying cell death pathways cancer research and immunology studies
Catalog No. 50-000-81816