Learn More
Meso Scale Discovery U-PLEX Human TSLP Assay, 96-well plate, 25 plates, dynamic range 0.20-10,100 pg/mL, for human sample types including Serum, Plasma, Cell Culture Supernatant
SDP
Supplier:  Meso Scale Discovery K151WHK4
Encompass
U-PLEX Human TSLP Assay, 96-well plate, 25 plates, dynamic range 0.20-10,100 pg/mL, for human sample types including Serum, Plasma, Cell Culture Supernatant
Catalog No. NC3791801
Only null left
Add to Cart
Request a Quote