Learn More
Meso Scale Discovery U-PLEX Human VCAM-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151H9K21
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit quantifies VCAM-1 in human serum plasma and cell culture supernatants using electrochemiluminescent detection It employs MSD GOLD Streptavidin Multi-Array 96-well plates and a VCAM-1 capture antibody A SULFO-TAG labeled detection antibody identifies the analyte Electricity applied to electrodes causes the SULFO-TAG to emit light measured by an MSD instrument The kit includes a calibrator and control for standard curve and validation It detects VCAM-1 in the range of 0 05-400 ng/mL
Catalog No. 50-000-80791