Learn More
Meso Scale Discovery U-PLEX Human VEGF-D ASY
SDP
Supplier:  Meso Scale Discovery K151C8K2
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay that measures VEGF-D levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less of sample and can be read on Meso Scale Discovery instrumentation platforms It includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82472