Learn More
Meso Scale Discovery U-PLEX Human vWF ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151C9K4
Encompass
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring VWF with a broad dynamic range of 1 56-1000 ng/mL The U-PLEX format allows for customization and multiplexing with other U-PLEX biomarker assays while using low sample volumes This assay can be used to aid in the diagnosis of von Willebrand disease and other conditions related to coagulation and platelet function
Catalog No. 50-000-82462
Add to Cart
Request a Quote