Learn More
Meso Scale Discovery U-PLEX Human vWF ASY
SDP
Supplier:  Meso Scale Discovery K151C9K1
Encompass
The Meso Scale Discovery U-PLEX Human VWF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring VWF with a broad dynamic range This multiplex assay allows simultaneous measurement of VWF along with other biomarkers It requires a low sample volume and provides high-quality data with low %CVs The U-PLEX platform enables efficient and flexible assay customization
Catalog No. 50-000-82031