Learn More
Meso Scale Discovery U-PLEX Human vWF ASY384-well
SDP
Supplier:  Meso Scale Discovery K251C9K2
Encompass
The Meso Scale Discovery U-PLEX Human VWF 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants The kit contains a VWF-specific antibody coated onto carbon electrodes in 384-well plates Samples are added followed by a VWF specific detection antibody conjugated to an electrochemiluminescent label Levels of VWF are determined using an MSD instrument by measuring emitted light upon electrochemical stimulation The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 100 ng/mL VWF in a 2 5% sample matrix
Catalog No. 50-000-81006
Add to Cart
Request a Quote