Learn More
Meso Scale Discovery U-PLEX Human YKL-40 ASYQP
SDP
Supplier:  Meso Scale Discovery K151VLK21
Encompass
The U-PLEX Human YKL-40 Assay (K151VLK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of YKL-40 biomarker in human samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes calibrators controls and detection reagents optimized for serum plasma and cell culture samples providing reliable results for research applications in inflammation and disease biomarker studies
Catalog No. 50-000-81957