Learn More
Meso Scale Discovery U-PLEX Mouse EPO ASY
SDP
Supplier:  Meso Scale Discovery K152VXK2
Encompass
The Meso Scale Discovery U-PLEX Mouse EPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in mouse serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of mouse EPO with an upper limit of quantification of 1000 pg/mL and a lower limit of quantification of 1 95 pg/mL The assay requires 50 L of sample and can be read on MSD s SECTOR Imager instruments This assay provides a rapid and robust method for measuring mouse EPO levels in research studies
Catalog No. 50-000-79367