Learn More
Meso Scale Discovery U-PLEX Mouse IFN- ASY
SDP
Supplier:  Meso Scale Discovery K152TTK1
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN gamma Assay is a multiplex immunoassay kit for measuring mouse interferon gamma (IFN-) in various samples It uses MSD s electrochemiluminescent detection technology and includes plates with capture antibodies detection antibodies calibrators controls buffers and reagents for 100 tests The assay is sensitive has a broad dynamic range and requires only 12 5 L per well It is suitable for monitoring IFN- in mouse models of infection autoimmunity cancer and other diseases
Catalog No. 50-000-78640