Learn More
Meso Scale Discovery U-PLEX Mouse IFN- ASYQP
SDP
Supplier:  Meso Scale Discovery K152W1K21
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is a multiplex immunoassay for measuring mouse interferon alpha in various samples It uses MSD s electrochemiluminescent detection for high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It detects mouse IFN alpha from 0 05 pg/mL to 10 000 pg/mL enabling efficient quantification of this antiviral cytokine
Catalog No. 50-000-79642