Learn More
Meso Scale Discovery U-PLEX Mouse IFN-B ASYQP
SDP
Supplier:  Meso Scale Discovery K152G0K22
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results in 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The dynamic range is 12 2-10 000 pg/mL requiring only 25 L of sample enabling high-throughput IFN beta profiling with minimal sample use
Catalog No. 50-000-82107