Learn More
Meso Scale Discovery U-PLEX Mouse IL-10 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152TZK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 10 (IL-10) in serum plasma cell culture supernatants and other biological samples This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay is for research use only and is not intended for diagnostic procedures The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent
Catalog No. 50-000-80435
Add to Cart
Request a Quote