Learn More
Meso Scale Discovery U-PLEX Mouse IL-12p70 ASY
SDP
Supplier:  Meso Scale Discovery K152UAK1
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 12 (IL-12) in serum plasma cell culture supernatants and other biological samples This assay uses MSD s proprietary SECTOR Imager to provide high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL This product enables reliable quantification of IL-12 for applications such as monitoring immune responses
Catalog No. 50-000-78983