Learn More
Meso Scale Discovery U-PLEX Mouse IL-15 ASY
SDP
Supplier:  Meso Scale Discovery K152URK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-15 Assay Kit is designed for the quantitative measurement of mouse interleukin 15 (IL-15) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range It uses MSD GOLD SULFO-TAG labels and is compatible with MSD instrument platforms The kit includes a plate coated with capture antibody detection antibody calibrator diluent wash buffer and read buffer
Catalog No. 50-000-79345