Learn More
Meso Scale Discovery U-PLEX Mouse IL-15 ASY384-well
SDP
Supplier:  Meso Scale Discovery K252URK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-15 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well plate format The kit includes a mouse IL-15 calibrator and pre-coated plates to enable the construction of a standard curve and rapid determination of sample concentrations This assay allows high-throughput analysis of IL-15 an important cytokine involved in lymphocyte development and proliferation
Catalog No. 50-000-81532
Add to Cart
Request a Quote