Learn More
Meso Scale Discovery U-PLEX Mouse IL-16 ASY
SDP
Supplier:  Meso Scale Discovery K152USK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-16 Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for generating a standard curve and measuring IL-16 levels accurately This assay allows efficient reproducible analysis of IL-16 an immunomodulatory cytokine involved in inflammatory and autoimmune responses
Catalog No. 50-000-79347