Learn More
Meso Scale Discovery U-PLEX Mouse IL-16 ASY384-well
SDP
Supplier:  Meso Scale Discovery K252USK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-16 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption It is available in a 384-well plate format includes all necessary reagents and can process hundreds of samples for the quantification of IL-16 This assay kit enables efficient and accurate measurement of the proinflammatory cytokine IL-16
Catalog No. 50-000-79325
Add to Cart
Request a Quote