Learn More
Meso Scale Discovery U-PLEX Mouse IL-16 ASYQP
SDP
Supplier:  Meso Scale Discovery K152USK21
Encompass
The U-PLEX Mouse IL-16 Assay (K152USK) is a highly sensitive immunoassay designed to quantify interleukin-16 levels in mouse biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering superior sensitivity and precision for cytokine detection in research applications The kit provides reliable quantitative measurement of IL-16 a key inflammatory mediator involved in immune cell recruitment and activation processes
Catalog No. 50-000-80442