Learn More
Meso Scale Discovery U-PLEX Mouse IL-1B ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152TUK4
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-1beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 1 beta (IL-1beta) in cell culture supernatants serum plasma and other biological samples This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available in a kit format that includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator
Catalog No. 50-000-79098
Only null left
Add to Cart
Request a Quote