Learn More
Meso Scale Discovery U-PLEX Mouse IL-1B ASYQP
SDP
Supplier:  Meso Scale Discovery K152TUK21
Encompass
The U-PLEX Mouse IL-1 Assay (K152TUK) is a highly sensitive electrochemiluminescence immunoassay designed to quantify interleukin-1 beta levels in mouse biological samples This multiplex-ready assay offers superior sensitivity and dynamic range compared to traditional ELISA methods enabling researchers to detect low-abundance cytokines in small sample volumes The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in inflammation and immunology research applications
Catalog No. 50-000-79102