Learn More
Meso Scale Discovery U-PLEX Mouse IL-21 ASY
SDP
Supplier:  Meso Scale Discovery K152WBK1
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-21 Assay Kit is a multiplex immunoassay for quantifying mouse interleukin 21 (IL-21) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for MSD instruments like the MESO QuickPlex SQ 120 The assay s sensitivity is 1 3 pg/mL with an upper quantification limit of 2500 pg/mL for mouse IL-21
Catalog No. 50-000-78683