Learn More
Meso Scale Discovery U-PLEX Mouse IL-21 ASY
SDP
Supplier:  Meso Scale Discovery K152WBK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-21 Assay Kit is a multiplex immunoassay that enables simultaneous detection and quantification of mouse interleukin 21 (IL-21) in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an MSD SULFO-TAGTM calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2-hour incubation
Catalog No. 50-000-78684