Learn More
Meso Scale Discovery U-PLEX Mouse IL-21 ASYQP
SDP
Supplier:  Meso Scale Discovery K152WBK22
Encompass
MSD s Mouse IL-21 U-PLEX assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-21 in mouse samples This multiplex-compatible assay uses SULFO-TAG labels and carbon electrodes for sensitive specific detection of IL-21 a cytokine involved in immune regulation and T-cell function The U-PLEX platform allows researchers to combine multiple analytes in a single well reducing sample volume requirements while maintaining high precision and reproducibility for mouse cytokine research applications
Catalog No. 50-000-81333