Learn More
Meso Scale Discovery U-PLEX Mouse IL-31 ASYQP
SDP
Supplier:  Meso Scale Discovery K152WEK22
Encompass
The MSD U-PLEX Mouse IL-31 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-31 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-31 a cytokine involved in inflammatory and allergic responses The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications studying immune responses skin disorders and inflammatory conditions in mouse models
Catalog No. 50-000-79055