Learn More
Meso Scale Discovery U-PLEX Mouse IL-5 ASY384-well
SDP
Supplier:  Meso Scale Discovery K252UOK2
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is a highly sensitive immunoassay that enables the quantitative measurement of mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides exceptional sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse IL-5
Catalog No. 50-000-79438
Only null left
Add to Cart
Request a Quote