Learn More
Meso Scale Discovery U-PLEX Mouse IL-6 ASYQP
SDP
Supplier:  Meso Scale Discovery K152TXK22
Encompass
The U-PLEX Mouse IL-6 Assay (K152TXK) is a highly sensitive electrochemiluminescence immunoassay designed to quantify interleukin-6 levels in mouse biological samples This multiplex-ready assay offers superior sensitivity and dynamic range compared to traditional ELISA methods enabling researchers to detect low-abundance cytokines in small sample volumes The assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms for reliable reproducible results in inflammation and immunology research applications
Catalog No. 50-000-79331