Learn More
Meso Scale Discovery U-PLEX Mouse IP-10 ASYQP
SDP
Supplier:  Meso Scale Discovery K152UFK22
Encompass
The U-PLEX Mouse IP-10 Assay (K152UFK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure IP-10 (CXCL10) levels in mouse biological samples This single-plex assay utilizes Meso Scale Discovery s advanced U-PLEX platform technology offering superior sensitivity and wide dynamic range for cytokine detection The kit includes pre-coated plates detection antibodies and calibration standards enabling researchers to accurately assess IP-10 concentrations in serum plasma and cell culture supernatants for inflammation and immune response studies
Catalog No. 50-000-78646