Learn More
Meso Scale Discovery U-PLEX Mouse MCP-5/CCL12 ASYQP
SDP
Supplier:  Meso Scale Discovery K152Y1K21
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection is 0 04 pg/mL
Catalog No. 50-000-81999