Learn More
Meso Scale Discovery U-PLEX Mouse MIP-2 ASY
SDP
Supplier:  Meso Scale Discovery K152XLK2
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-2 Assay is a multiplex immunoassay kit for measuring mouse macrophage inflammatory protein 2 (MIP-2) in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for generating a standard curve to quantify MIP-2 concentrations enabling rapid and accurate measurement of this pro-inflammatory chemokine in mouse samples
Catalog No. 50-000-81870