Learn More
Meso Scale Discovery U-PLEX Mouse NGAL/LCN2 ASYQP
SDP
Supplier:  Meso Scale Discovery K152Z1K22
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in mouse biological samples such as serum plasma and cell culture supernatants This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other cytokines The assay requires a small sample volume of 50 L and provides results in under 3 hours This product can aid in the study of acute kidney injury and other inflammatory conditions where NGAL is a biomarker
Catalog No. 50-000-82117