Learn More
Meso Scale Discovery U-PLEX Mouse PYY total ASYQP
SDP
Supplier:  Meso Scale Discovery K1526BK21
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for measuring total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective measurement with high specificity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software
Catalog No. 50-000-81495