Learn More
Meso Scale Discovery U-PLEX Mouse RANTES ASY
SDP
Supplier:  Meso Scale Discovery K152A2K2
Encompass
The Meso Scale Discovery U-PLEX Mouse RANTES Assay is a multiplex immunoassay kit for measuring RANTES in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies calibrators and controls Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software providing a rapid reproducible method for measuring mouse RANTES
Catalog No. 50-000-81498