Learn More
Meso Scale Discovery U-PLEX Mouse TARC ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152B0K4
Encompass
The Meso Scale Discovery U-PLEX Mouse TARC Assay is a multiplex immunoassay kit for measuring mouse Thymus and Activation Regulated Chemokine (TARC/CCL17) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a range of 0 3 to 10 000 pg/mL Requiring 50 L of sample it is compatible with MSD platforms like MESO QuickPlex SQ 120 SECTOR Imager 2400 and SECTOR S 600 offering a simple rapid and reproducible method for measuring mouse TARC
Catalog No. 50-000-82104
Add to Cart
Request a Quote