Learn More
Meso Scale Discovery U-PLEX Mouse TARC ASYQP
SDP
Supplier:  Meso Scale Discovery K152B0K22
Encompass
The U-PLEX Mouse TARC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TARC (thymus and activation regulated chemokine) in mouse serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period The lower limit of detection is 1 3 pg/mL
Catalog No. 50-000-82209