Learn More
Meso Scale Discovery U-PLEX Mouse TARC ASYQP
SDP
Supplier:  Meso Scale Discovery K152B0K21
Encompass
The Meso Scale Discovery U-PLEX Mouse TARC Assay is a multiplex immunoassay kit for measuring mouse Thymus and Activation Regulated Chemokine (TARC/CCL17) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibody detection antibody calibrator and assay diluent It quantifies mouse TARC with a detection range of 1 3 to 10 000 pg/mL offering a simple rapid and cost-effective analysis method
Catalog No. 50-000-82359